Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium hydroxide
Sodium hydroxide 1/2.8 mol/l (1/2.8 N) in aqueous solution
Sodium hydroxide 1/2.8 mol/l (1/2.8 N) in aqueous solution
Sodium hydroxide 1/2.8 mol/l (1/2.8 N) in aqueous solution
PRLSMC310887.6010
:
PROLABO CMP
:
Sodium hydroxide 1/2.8 mol/l (1/2.8 N) in aqueous solution
PRLSMC310887.6010
:
PROLABO CMP
:
Pk:
10 L
Seq:
5214
:
:
40 g/mol
:
∼105 °C (1013 hPa)
:
1.02 g/cm³ (25 °C)
:
Ambient
:
215-185-5
:
1824
:
8,III
:
GHS07
:
1310-73-2
:
NaOH
:
MFCD00003548