Order Entry
France
ContactUsLinkComponent
Hydrochloric acid 6 mol/l (6 N)
Hydrochloric acid 6 mol/l (6 N)
Catalog # VWRS83910.360
Supplier:  VWR Collection
CAS Number:  
Hydrochloric acid 6 mol/l (6 N)
Catalog # VWRS83910.360
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pack type:
    Plastic container
  • Pk:
    5 L
  • Seq:
    5190
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Hydrochloric acid solution, Muriatic acid
 :
 :36.46 g/mol
EINECS:231-595-7
UN:1789
 :GHS07
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324
Density:1.06 g/cm³ (20 °C)
ADR:8 II