Order Entry
France
ContactUsLinkComponent
Dithiothreitol (DTT, Cleland's reagent) 0.1 mol/l in aqueous solution
Dithiothreitol (DTT, Cleland's reagent) 0.1 mol/l in aqueous solution
Catalog # TREV4145-100-02
Supplier:  TREVIGEN
CAS Number:  
undefined
Dithiothreitol (DTT, Cleland's reagent) 0.1 mol/l in aqueous solution
Catalog # TREV4145-100-02
Supplier:  TREVIGEN
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Seq:
    0075

Specifications

About this item

 :
 :154.25 g/mol
EINECS:222-468-7
CAS Number:3483-12-3
 :27565-41-9
Formula:C₄H₁₀O₂S₂
MDL Number:MFCD00004877