Order Entry
France
ContactUsLinkComponent
Disodium oxalate 0.05 mol/l (0.1 N) in aqueous solution
Disodium oxalate 0.05 mol/l (0.1 N) in aqueous solution
Catalog # PRLSMC311855.0500
Supplier:  PROLABO CMP
CAS Number:  
Disodium oxalate 0.05 mol/l (0.1 N) in aqueous solution
Catalog # PRLSMC311855.0500
Supplier:  PROLABO CMP
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :134 g/mol
Storage Temperature:Ambient
EINECS:200-550-3
CAS Number:62-76-0
Formula:Na₂C₂O₄
MDL Number:MFCD00012465