DL-Tryptophan ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:54-12-6
:
Formula:C₁₁H₁₂N₂O₂
MDL Number:MFCD00064339
:204.23 g/mol
Melting Pt:∼ 280 °C (dec.)
Density:1.349 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-194-9
Related Information
Frequently Bought Together