


Allyl isothiocyanate 94% stabilised
:57-06-7
:
:C₄H₅NS
:MFCD00004822
:99.16 g/mol
:13,00292
:151…153 °C (760 mmHg)
:–80 °C
:1.0175 g/cm³
:46 °C
:Refrigerator
:1545
:6.1,II
:Stabilised with α-tocopherol 0.01%
:200-309-2
:GHS06
:GHS09
:GHS02
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5300 to 1.5330 (20°C, 589 nm)
- GC:≥92 %
- Stabilizer:0.01 % alpha-Tocopherol
- Relative density:(20/4°C) 1.017 to 1.022
Frequently Bought Together