


1,1-Dichloroethylene stabilised for synthesis, Sigma-Aldrich®
: Merck
:75-35-4
:
:C₂H₂Cl₂
:MFCD00011653
:12,10134
:31.7 °C (1013 hPa)
:–122 °C
:1.21 g/cm³ (20 °C)
:–25 °C
:Refrigerator
:3,I
:M-Clarity™ quality level = MQ200
:Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
:96.94 g/mol
:200-864-0
:1303
:GHS02
:GHS08
:GHS07
- Solubility:2.5 g/l (25 °C)
- Melting Point:-122 °C
- Molar Mass:96.94 g/mol
- Boiling Point:31.2 - 31.7 °C (1013 hPa)
- Vapor Pressure:665 hPa (20 °C)
- Flash Point:-25 °C
- Refractive Index:1.4254 (20 °C, 589 nm)
- Explosion Limit:8.4 - 16.5 %(V)
- Density:1.21 g/cm3 (20 °C)
- Ignition Point:520 °C
- Hazard Class:3
- Hazard Class Description:Flammable liquid
Frequently Bought Together