DL-Lithium lactate
Catalog # MOLE43935171-100G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:96.01 g/mol
Melting Pt:300 °C
Storage Temperature:Ambient
EINECS:212-761-8
CAS Number:867-55-0
Formula:H₃CCH(OH)COOLi
MDL Number:MFCD00065399