DL-Lithium lactate, powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:867-55-0
:
Formula:H₃CCH(OH)COOLi
MDL Number:MFCD00065399
:96.01 g/mol
Melting Pt:300 °C
Storage Temperature:Ambient
EINECS:212-761-8
Specification Test Results
- Identity Test:Passes
- FTIR:Conforms to Standard
Specification Test Results
Frequently Bought Together