Chloroform 99+% stabilised, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:67-66-3
:
Formula:CHCl₃
MDL Number:MFCD00000826
:119.38 g/mol
Merck Index:13,02160
Boiling Pt:61.7 °C (1013 hPa)
Melting Pt:–63 °C
Density:1.48 g/cm³
Storage Temperature:Ambient
UN:1888
ADR:6.1,III
:Stabilised with ethanol
EINECS:200-663-8
REACH:05-2114614906-45
:GHS06
:GHS08
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Water:≤0.02 % (K.F.)
- GC:≥99.0 % (excluding stabilizer)
- Color scale:≤25 APHA
- Stabilizer:0.5 to 1.0 % ethanol
- Residue after evaporation:≤0.002 %
- Ammonium (NH4):≤0.002 %
- Calcium (Ca):≤0.005 %
- Iron (Fe):≤0.001 %
- Heavy metals (as Pb):≤5 ppm
- Insoluble matter:≤0.005 %
Specification Test Results
Frequently Bought Together