Order Entry
France
ContactUsLinkComponent
Dichloromethane 99+% stabilised, extra pure
Health
Irritant
Dichloromethane 99+% stabilised, extra pure
Certificates
undefined
Health
Irritant
Dichloromethane 99+% stabilised, extra pure

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.1 % (K.F.)
  • GC:
    ≥99.0 %
  • Stabilizer:
    Approx. 0.2 % Ethanol
  • Residue after evaporation:
    ≤0.05 %

Specification Test Results

Related Information

Frequently Bought Together