Order Entry
France
Orders LinkContactUsLinkComponent
Tween® 20 (Polysorbate), TECHNICAL

Tween® 20 (Polysorbate), TECHNICAL

Plastic bottle, 1 L
Catalog # 28829.296
Supplier:   VWR Chemicals
Tween® 20 (Polysorbate), TECHNICAL
Catalog # 28829.296
Supplier:  VWR Chemicals
CAS Number:  9005-64-5
Tween® 20 (Polysorbate), TECHNICAL
Tween® 20 (Polysorbate), TECHNICAL

Plastic bottle, 1 L

CAS Number:  9005-64-5
Price
53,40 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Polyoxyethylene sorbitan monolaurate
  • CAS Number:9005-64-5
  • Formula:C₅₈H₁₁₄O₂₆
  • MDL Number:MFCD00165986
  • Molecular Weight:1227.53 g/mol
  • Density:1.1 g/cm³ (20 °C)
  • Flash Pt:149 °C (closed cup)
  • Storage Temperature:Ambient
Specifications

Specification Test Results

  • Hydroxyl value:
    90 - 110
  • Appearance:
    Yellow to brown-yellow

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.