Order Entry
France
ContactUsLinkComponent
Benzalkonium chloride 50% in aqueous solution
Corrosive
Irritant
Environment
Benzalkonium chloride 50% in aqueous solution
Catalog # MOLE29089235-500ML
Supplier:  Molekula
CAS Number:  
Corrosive
Irritant
Environment
Benzalkonium chloride 50% in aqueous solution
Catalog # MOLE29089235-500ML
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
Storage Temperature:Ambient
EINECS:264-151-6
UN:3265
ADR:8,III
 :GHS05
 :GHS07
 :GHS09
CAS Number:63449-41-2
 :8001-54-5
Formula:C₆H₅CH₂N(CH₃)₂RCl (R=C₈H₁₇ to C₁₈H₃₇)
MDL Number:MFCD00145757