Order Entry
France
ContactUsLinkComponent
VWR® Acryl/Bis™ Acrylamine/Bis-Acrylamine
VWR® Acryl/Bis™ Acrylamine/Bis-Acrylamine
Catalog # M157-500ML
Supplier:  VWR Chemicals
undefined
VWR® Acryl/Bis™ Acrylamine/Bis-Acrylamine
Catalog # M157-500ML
Supplier:  VWR Chemicals
Specifications
  • Description:
    Acryl/Bis™
  • Pk:
    500 mL
Specifications
About this item
A 40% (w/v) solution of ultra pure acrylamide (38,96%) and bis-acrylamide (1,04%) at a final ratio of 37.5:1.
  • Ultra-pure acrylamide powders (>99,9%)
  • High solubility, producing haze-free solutions
  • Liquid stable solutions eliminate handling hazardous powdered acrylamide
  • Reproducible and consistent results
VWR® offers an extensive line of acrylamide and bis-acrylamide pre-weighed powder blends, pre-mixed stock solutions and ready-to-use solutions for customised polyacrylamide gel electrophoresis of proteins. Ultra-pure (>99,9%) acrylamide and bis-acrylamide powders provide the flexibility to prepare solutions having concentrations and ratios for all electrophoresis applications. Liquid blends minimise the handling of neurotoxic acrylamide.