Order Entry
France
ContactUsLinkComponent
L(+)-Arabinose
L(+)-Arabinose
Catalog # ROTH3138.1
Supplier:  Roth Carl
CAS Number:  
undefined
L(+)-Arabinose
Catalog # ROTH3138.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :150.13 g/mol
Boiling Pt:331 °C (1013 hPa)
Melting Pt:158…160 °C
Density:1.585 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:226-214-6
CAS Number:5328-37-0
 :87-72-9
Formula:C₅H₁₀O₅
MDL Number:MFCD00067709