Order Entry
France
ContactUsLinkComponent
Western blot kits, TrueBlot®
Western blot kits, TrueBlot®
Certificates
undefined
Western blot kits, TrueBlot®

Specifications

  • Storage temperature:
    −20 °C
  • Concentration:
    1,0 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Wet ice

Specifications

Related Information

Frequently Bought Together