Triammonium citrate in aqueous solution
Catalog # PRLSMC311006.1000
Supplier: PROLABO CMP
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:243.22 g/mol
Melting Pt:29 to 34 °C
CAS Number:3458-72-8
Formula:C₆H₁₇N₃O₇
MDL Number:MFCD00036406