



Petroleum spirit, 40…60 °C, AnalaR NORMAPUR® ACS analytical reagent (max. 0.01% aromatic hydrocarbons)
:64742-49-0
:64742-82-1
:MFCD00081849
:40…60 °C (1013 hPa)
:0.645…0.665 g/cm³ (20 °C)
:–40 °C
:Ambient
:265-151-9
:1268
:3,II
:01-2119474679-18
:GHS02
:GHS08
:GHS07
:GHS09
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Acidity:Max. 0.0002 meq/g
- Bromine value:Max. 1
- Colouration:Max. 10 APHA
- Boiling range (40-60°C) (Vol %):Min. 90%
- Substances coloured by H2SO4:Max. 150 APHA
- Benzene:Max. 100 ppm
- Evaporation residue:Max. 10 ppm
- Total S (as SO4):Max. 50 ppm
- Water:Max. 100 ppm
- Al (Aluminium):Max. 0.2 ppm
- Ba (Barium):Max. 0.1 ppm
- Ca (Calcium):Max. 0.5 ppm
- Cd (Cadmium):Max. 0.05 ppm
- Co (Cobalt):Max. 0.02 ppm
- Cr (Chromium):Max. 0.02 ppm
- Cu (Copper):Max. 0.02 ppm
- Fe (Iron):Max. 0.1 ppm
- K (Potassium):Max. 0.3 ppm
- Mg (Magnesium):Max. 0.1 ppm
- Mn (Manganese):Max. 0.02 ppm
- Na (Sodium):Max. 1 ppm
- Ni (Nickel):Max. 0.02 ppm
- Pb (Lead):Max. 0.1 ppm
- Sn (Tin):Max. 0.1 ppm
- Sr (Strontium):Max. 0.03 ppm
- Zn (Zinc):Max. 0.1 ppm
- Conforms to ACS:Passes test
1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249) 225L Metal drum UN (ZPACE8010050) 30L Metal drum for liquids UN X (ZPACK12124125) 5L Aluminium bottle for liquids DIN 45 UN X (ZPACK8000430)
Frequently Bought Together