Gold ≥99.9985% (metals basis), wire, Premion®, Ø 2.0 mm (0.08 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-57-5
:
Formula:Au
MDL Number:MFCD00003436
:196.97 g/mol
Merck Index:12,04526
Boiling Pt:2800 °C (1013 hPa)
Melting Pt:1063 °C
Density:19.3 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-165-9
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together