Arsenic standard solution, 10000 mg/L As in 5% nitric acid (from As) standard for AAS and ICP
Catalog # 634-0139
Supplier: JOBIN YVON EMISSION
CAS Number:
Specifications
- Concentration:10000 mg/L
- Matrix:5% HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Arsenic
- Symbol:As
Specifications
About this item
Certifications: Traceable to SRM from NIST, tested in ISO 17034 / ISO 17025 accredited laboratory.