Charcoal activated, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7440-44-0
Formula:C
MDL Number:MFCD00133992
:12.01 g/mol
Merck Index:13,01818
Melting Pt:3550 °C
Density:2 g/cm³ (20 °C)
EINECS:231-153-3
UN:1362
ADR:4.2,III
:M-Clarity™ quality level = MQ100
REACH:01-2119488894-16
Frequently Bought Together