Supplements for renal epithelial cell growth medium 2
Supplier: PROMOCELL
Certificates
About this item
PromoCell Renal Epithelial Cell Growth Medium 2 is an optimised low serum medium that contains epinephrine. The medium consists of Basal Medium plus SupplementMix. Supplements are also available to purchase separately, either in separate vials (SupplementPack) or as a pre-prepared mix (SupplementMix).
Frequently Bought Together