Order Entry
France
ContactUsLinkComponent
Human Uterine smooth muscle cells (HUtSMC)
Human Uterine smooth muscle cells (HUtSMC)
Catalog # C-14059
Supplier:  PROMOCELL
CAS Number:  
undefined
Human Uterine smooth muscle cells (HUtSMC)
Catalog # C-14059
Supplier:  PROMOCELL
CAS Number:  

Specifications

  • Cell Format:
    Pelleted
  • Pk:
    2 mL
  • Description:
    Primary Human Uterine Smooth Muscle Cells (HUtSMC), pellet, 1×106 cells in RNAlater®

Specifications

About this item

Primary Human Uterine Smooth Muscle Cells (HUtSMC) are isolated from the myometrium (the middle layer of the uterine wall) and stain positive for smooth muscle α-actin. During pregnancy, hormones and growth factors cause an extensive hypertrophy of uterine smooth muscle cells. This and other morphological and biochemical effects on the uterus can be studied in vitro using HUtSMC.

Applications include: Organ specific drug testing, cell-extracellular matrix interaction studies, physiological and biochemical studies, oncology, pathological studies (e.g. hypertension).

Ordering information: Human Uterine Smooth Muscle Cells can be ordered as cryopreserved or proliferating cells. Cell pellets stored in RNAlater® are also available.