Order Entry
France
ContactUsLinkComponent
Human Umbilical artery endothelial cells (HUAEC)
Human Umbilical artery endothelial cells (HUAEC)
Catalog # C-14013
Supplier:  PROMOCELL
CAS Number:  
undefined
Human Umbilical artery endothelial cells (HUAEC)
Catalog # C-14013
Supplier:  PROMOCELL
CAS Number:  

Specifications

  • Cell Format:
    Pelleted
  • Pk:
    2 mL
  • Description:
    Primary Human Umbilical Artery Endothelial Cells (HUAEC) pellet, 1×106 cells in RNAlater®

Specifications

About this item

Primary Human Umbilical Artery Endothelial Cells (HUAEC) are isolated from the arteries of the umbilical cord and are often used for in vitro studies on atherosclerosis but also for endothelial cell-dependent lymphocyte activation and chemoattractant activity experiments.

  • Von Willebrand factor (vWF) positive
  • CD31 positive, Dil-Ac-LDL uptake positive
  • Smooth muscle alpha-actin negative

Applications include: Angiogenesis research, atherosclerosis research, oncology, drug uptake studies, cell-cell junction investigation, inflammation studies.

Ordering information: Human Umbilical Artery Endothelial Cells can be ordered as cryopreserved or proliferating cells. Cell pellets stored in RNAlater® are also available.