Order Entry
France
ContactUsLinkComponent
Human small airway epithelial cells (HSAEpC)
Human small airway epithelial cells (HSAEpC)
Catalog # C-14065
Supplier:  PROMOCELL
CAS Number:  
undefined
Human small airway epithelial cells (HSAEpC)
Catalog # C-14065
Supplier:  PROMOCELL
CAS Number:  

Specifications

  • Cell Format:
    Pelleted
  • Pk:
    2 mL
  • Description:
    Primary Human Small Airway Epithelial Cells (HSAEpC) pellet, 1×106 cells in RNAlater®

Specifications

About this item

Primary Human Small Airway Epithelial Cells (HSAEpC) are isolated from the distal portion of the human respiratory tract in the 1 mm bronchiole area. They stain positive for cytokeratin.

The distal respiratory tract mainly consists of pulmonary alveoli, which are spherical outcroppings of the respiratory bronchioles and are the primary sites of gas exchange with the blood. HSAEpC are qualified for functional studies to investigate disorders such as asthma or pulmonary inflammation.

Applications include: Drug uptake studies, toxicological research, wound healing research, oncology, cell differentiation studies, respiratory disease research.

Ordering information: Human Small Airway Epithelial Cells can be ordered as cryopreserved or proliferating cells. Cell pellets stored in RNAlater® are also available.