Order Entry
France
ContactUsLinkComponent
Human Umbilical artery smooth muscle cells (HUASMC)
Human Umbilical artery smooth muscle cells (HUASMC)
Catalog # C-14050
Supplier:  PROMOCELL
CAS Number:  
undefined
Human Umbilical artery smooth muscle cells (HUASMC)
Catalog # C-14050
Supplier:  PROMOCELL
CAS Number:  

Specifications

  • Cell Format:
    Pelleted
  • Pk:
    2 mL
  • Description:
    Primary Human Umbilical Artery Smooth Muscle Cells (HUASMC) pellet, 1×106 cells in RNAlater®

Specifications

About this item

Primary Human Umbilical Artery Smooth Muscle Cells (HUASMC) are isolated from the arteries of the umbilical cord and stain positive for smooth muscle α-actin. Physiological and biochemical processes of vascular diseases can be analysed with HUASMC in culture.

Applications include: Organ specific drug testing, cell-extracellular matrix interaction studies, physiological and biochemical studies, oncology, pathological studies (e.g. hypertension).

Ordering information: Human Umbilical Artery Smooth Muscle Cells can be ordered as cryopreserved or proliferating cells. Cell pellets stored in RNAlater® are also available.