Order Entry
France
ContactUsLinkComponent
Human aortic adventitial fibroblasts (HAoAF)
Human aortic adventitial fibroblasts (HAoAF)
Catalog # C-14037
Supplier:  PROMOCELL
CAS Number:  
undefined
Human aortic adventitial fibroblasts (HAoAF)
Catalog # C-14037
Supplier:  PROMOCELL
CAS Number:  

Specifications

  • Cell Format:
    Pelleted
  • Pk:
    2 mL
  • Description:
    Human Aortic Adventitial Fibroblasts (HAoAF) pellet, 1×106 cells in RNAlater®

Specifications

About this item

Primary Human Aortic Adventitial Fibroblasts (HAoAF) are isolated from human aortic adventitial tissue. HAoAF can be used for studies concerning vascular diseases e.g. neointimal formation, vascular remodeling, atherosclerosis, and hypertension. They are also often used for the examination of fibroblast disorders like fibrosis or other diseases linked to either imperfect or excessive accumulation of fibroblasts.

Applications include: Wound healing research, tissue engineering research, regeneration studies, toxicological research, oncology, epithelial-mesenchymal interaction studies.

Ordering information: Human Aortic Adventitial Fibroblasts can be ordered as cryopreserved or proliferating cells. Cell pellets stored in RNAlater® are also available.