Anti-IgG Horse Polyclonal Antibody
IMMRHR-003-CBIO
- Antibody type:Secondary
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Horse
- Isotype:
- Reactivity:
- Environmentally Preferable:
- Form:Liquid
- Concentration:1.5 mg/ml
- Shipping temperature:2-8 °C
- Pack type:Glass vial
- Pk:1 mg
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Horse
Isotype:
Reactivity: