Order Entry
France
ContactUsLinkComponent
Dulbecco's phosphate-buffered saline (DPBS)
Dulbecco's phosphate-buffered saline (DPBS)
Catalog # C-40230
Supplier:  PROMOCELL
CAS Number:  
undefined
Dulbecco's phosphate-buffered saline (DPBS)
Catalog # C-40230
Supplier:  PROMOCELL
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Description:
    Dulbecco’s PBS
  • Size:
    500 mL

Specifications

About this item

Dulbecco's PBS solution for use in tissue culture, cell isolation procedures and molecular biology.

  • Ready to use buffers
  • Storage at 4 to 8 °C

Routinely tested for absence of cytotoxicity, and physical parameters such as osmolality and pH level.