Order Entry
France
ContactUsLinkComponent
Human accutase solution
Human accutase solution
Catalog # C-41310
Supplier:  PROMOCELL
CAS Number:  
undefined
Human accutase solution
Catalog # C-41310
Supplier:  PROMOCELL
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Species:
    Human
  • Enzyme name:
    Accutase solution

Specifications

About this item

Accutase is a sterile filtered solution specially developed for gentle and effective cell dissociation.

  • Ready to use
  • No stabilisers or other proteins have been added
  • Accutase supplied in Dulbecco's PBS containing 0,5 mM EDTA

Accutase solution is a unique mixture of proteolytic and collagenolytic enzymes from an invertebrate species. Accutase has been ultra-filtered before lyophilisation and contains only purified enzymes.