Dulbecco's phosphate-buffered saline (DPBS) without Ca²⁺ and Mg²⁺
C-40232
:
- Pk:500 mL
- Description:Dulbecco’s PBS, without Ca2+/Mg2+
These dulbecco's PBS solution are used in tissue culture, cell isolation procedures and molecular biology.
- Ready to use buffers
- Storage at 4 to 8 °C
Routinely tested for absence of cytotoxicity, and physical parameters such as osmolality and pH level.