Rat recombinant nNOS
Catalog # ENZOALX2018540010
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Species:Rat
- Seq:02
- Protein synonyms:bNOS|Nitric oxide synthase (neuronal)|NOS I|Brain NOS|Neuronal NOS
- Protein/peptide name:nNOS
- Purity:>90% (SDS-PAGE)
- Molecular weight:~160kDa/subunit; homodimer
- Formulation:Liquid. In 50 mM TRIS, pH 7,4, containing 0,1 mM EDTA, 10% glycerol, 100 mM NaCl, 0,1 mM DTT, 10 µM tetrahydrobiopterin.
Specifications
About this item
Produced in E. coli. Untagged rat nNOS.