Order Entry
France
ContactUsLinkComponent
Human recombinant SUMO2 (from E. coli), His-tag
Human recombinant SUMO2 (from E. coli), His-tag
Catalog # ENZOBMLUW92050500
Supplier:  Enzo Life Sciences
CAS Number:  
Human recombinant SUMO2 (from E. coli), His-tag
Catalog # ENZOBMLUW92050500
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    500 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Tag sequence:
    HIS Tag
  • Protein/peptide name:
    SUMO2

Specifications

About this item

Produced in E. coli. Human SUMO-2 is fused to a His-tag.

Mature protein for functional studies.