Order Entry
France
ContactUsLinkComponent
Di-Ubiquitin (linear) (from E. coli)
Di-Ubiquitin (linear) (from E. coli)
  ENZOBMLUW07750100
 :  
Di-Ubiquitin (linear) (from E. coli)
  ENZOBMLUW07750100
 :  

 

  • Pk:
    100 µG
  • Source:
    E. coli
  • Seq:
    02
  • Protein synonyms:
    UB2
  • Protein/peptide name:
    Di-Ubiquitin (linear)
  • Purity:
    >95% (SDS-PAGE)
  • Concentration:
    1 mg/ml
  • Formulation:
    Liquid. In 50 mM TRIS, pH 8,0, containing 50 mM sodium chloride.
  • Purification:
    Purified by multistep chromatography

 

 

Linear polyubiquitin chains are useful tools for investigating, amongst other things, the specificity and reactivity of deubiquitinylating enzymes (DUBs) and the recognition and interaction of linear polyubiquitin modified proteins with ubiquitin binding domain (UBDs) containing proteins.