Human recombinant NEDD8 (from E. coli), Biotin
Catalog # ENZOBMLUW05600100
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:100 µG
- Conjugation:Biotin
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Reconstitution instructions:In order to ensure full solubility it is recommended that the material be dissolved in 5 µl dimethyl sulphoxide (DMSO) prior to dilution with the buffer of choice
- Protein/peptide name:NEDD8
- Formulation:Lyophilised solid (from 1:1 acetonitrile:water)
Specifications
About this item
Produced in E. coli.