Order Entry
France
ContactUsLinkComponent
Human recombinant p300 (catalytic domain) (from E. coli)
Human recombinant p300 (catalytic domain) (from E. coli)
Catalog # ENZOBMLSE4510100
Supplier:  Enzo Life Sciences
CAS Number:  
Human recombinant p300 (catalytic domain) (from E. coli)
Catalog # ENZOBMLSE4510100
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Protein/peptide name:
    p300 (catalytic domain)
  • Purity:
    >75% (SDS-PAGE)
  • Formulation:
    Liquid. In 50 mM TRIS/HCl, pH 8,0, containing 0,1mM EDTA, 10% glycerol.

Specifications

About this item

Produced in E. coli. Catalytic domain (aa 1284-1673) of human p300.