Order Entry
France
ContactUsLinkComponent
Human recombinant MMP-10 (catalytic domain) (from E. coli)
Human recombinant MMP-10 (catalytic domain) (from E. coli)
Catalog # ENZOBMLSE3290010
Supplier:  Enzo Life Sciences
CAS Number:  
Human recombinant MMP-10 (catalytic domain) (from E. coli)
Catalog # ENZOBMLSE3290010
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Protein/peptide name:
    MMP-10 (catalytic domain)

Specifications

About this item

Produced in E. coli. Active Matrix Metalloproteinase-10 (MMP-10, stromelysin-2, transin-2) catalytic domain from human cDNA.

The enzyme consists of the catalytic domain of human MMP-10 (Phe99-Glu271, NM_2425) with a C-terminal purification tag. This comprises an active form of MMP-10 which lacks the C-terminal hemopexin domain.

MMPs lacking this domain cannot cleave native collagens; however, activity toward other targets such as gelatin, casein, or peptide substrates is unaffected.