Order Entry
France
ContactUsLinkComponent
Human recombinant Calmodulin (from E. coli)
Human recombinant Calmodulin (from E. coli)
  ENZOBMLSE3250001
 :  
undefined
Human recombinant Calmodulin (from E. coli)
  ENZOBMLSE3250001
 :  

 

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Biological activity:
    Elicits Ca2+-dependent activation of calcineurin (Ca2+/calmodulin-dependent Ser/Thr protein phosphatase 2B, PP-2B) with the same potency and to the same maximum extent as purified native calmodulin.
  • Reconstitution instructions:
    Reconstitute in water or an appropriate buffer.
  • Protein/peptide name:
    Calmodulin
  • Purity:
    >98% (SDS-PAGE)
  • Formulation:
    Lyophilised. Salt-free.

 

 

Produced in E. coli.