Human recombinant Fc:CD40L (soluble)
Catalog # ENZOALX522076C010
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Species:Human
- Seq:02
- Biological activity:Stimulates the proliferation of human dendritic cells. Note: Does not require an enhancer.
- Reconstitution instructions:Reconstitute with 100 µl sterile water. Further dilutions should be made with medium containing 5% fetal calf serum or a carrier protein.
- Protein/peptide name:Fc:CD40L (soluble)
- Purity:>90% (SDS-PAGE)
- Concentration:0,1 mg/ml after reconstitution
- Formulation:Lyophilised. Contains PBS.
Specifications
About this item
Produced in HEK 293 cells. The extracellular domain of human CD40L (CD154) (aa 116-261) is fused at the N-terminus to the Fc portion of human IgG1 and a linker peptide (20 aa).