Order Entry
France
ContactUsLinkComponent
Human recombinant IL-1beta (soluble) (fromE. coli)
Human recombinant IL-1beta (soluble) (fromE. coli)
Supplier:  ENZO LIFE SCIENCES
Certificates
Human recombinant IL-1beta (soluble) (fromE. coli)
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Biological activity:
    Activates the NF-κB pathway.
  • Reconstitution instructions:
    Reconstitute with 100 µl sterile water.
  • Protein/peptide name:
    IL-1beta (soluble)
  • Purity:
    >90% (SDS-PAGE)
  • Concentration:
    0,1 mg/ml after reconstitution
  • Formulation:
    Lyophilised. Contains PBS.

Specifications

Frequently Bought Together