Order Entry
France
ContactUsLinkComponent
 
Mouse recombinant GITR:Fc
Catalog # ENZOALX522024C050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Mouse recombinant GITR:Fc
Catalog # ENZOALX522024C050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Seq:
    02
  • Reconstitution instructions:
    Reconstitute with 50 µl sterile water. Further dilutions should be made with medium containing 5% fetal calf serum or a carrier protein.
  • Protein/peptide name:
    GITR:Fc
  • Purity:
    >95% (SDS-PAGE)
  • Concentration:
    1 mg/ml after reconstitution
  • Formulation:
    Lyophilised. Contains PBS.

Specifications

About this item

Produced in HEK 293 cells. The cysteine-rich region of mouse GITR (aa 20-152) is fused to the Fc portion of human IgG1.