Order Entry
France
ContactUsLinkComponent
Human recombinant Fas:Fc
Human recombinant Fas:Fc
Catalog # ENZOALX522002C050
Supplier:  Enzo Life Sciences
CAS Number:  
Human recombinant Fas:Fc
Catalog # ENZOALX522002C050
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Seq:
    02
  • Biological activity:
    Inhibits the activity of human and mouse soluble Fas (sFasL) and thereby sFasL-mediated lysis of Fas sensitive cells in a concentration range of 20 - 100 µg/ml.
  • Reconstitution instructions:
    Reconstitute with 50 µl sterile water. Further dilutions can be made with medium containing 5% fetal calf serum or a carrier protein.
  • Protein/peptide name:
    Fas:Fc
  • Purity:
    >95% (SDS-PAGE)
  • Concentration:
    1 mg/ml after reconstitution
  • Formulation:
    Lyophilised. Contains PBS.

Specifications

About this item

Produced in HEK 293 cells. The extracellular domain of human Fas (CD95; APO-1) (aa 7-154) is fused to the Fc portion of human IgG1.