Order Entry
France
ContactUsLinkComponent
Human recombinant beta-Nerve Growth Factor (from E. coli)
Human recombinant beta-Nerve Growth Factor (from E. coli)
Catalog # ENZOALX521005C005
Supplier:  Enzo Life Sciences
CAS Number:  
Human recombinant beta-Nerve Growth Factor (from E. coli)
Catalog # ENZOALX521005C005
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    5 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Biological activity:
    Neurotrophic activity was bioassayed using rat pheochromocytoma PC12 cell cultures over a period of 5 - 14 days. Activity is in the range  of 1 - 50 ng/ml.
  • Reconstitution instructions:
    Reconstitue in water or DMEM to a final concentration of at least 10 µg/ ml and store at -80°C before use.
  • Protein/peptide name:
    beta-Nerve Growth Factor
  • Purity:
    >98% (HPLC)
  • Formulation:
    Lyophilised. No additives.

Specifications

About this item

Produced in E. coli. Dimer of two identical subunits (119 aa) linked together by very strong hydrophobic interactions.

Ordering information: Quantity: 5 µg (determined by amino acid analysis).