Recombinant DnaJ (from E. coli), HIS tag
Produced in E. coli. E. coli DnaJ is fused at the N-terminus to a His-tag.
- Protein/peptide type:Recombinant
- Source:E. coli
- Tag sequence:HIS Tag
- Protein/peptide name:DnaJ
- Purity:>90% (SDS-PAGE; Western blot)
- Formulation:Liquid, in 25 mM HEPES, pH 7,5, containing 200 mM potassium chloride, 10% glycerol, and 5 mM DTT
- Purification:Purified by multi-step chromatography
Frequently Bought Together