GFP-Certified™ FluoForte™ Reagent
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Calcium dye
The GFP-Certified™ FluoForte™ Reagent is the brightest and most sensitive fluorescent calcium indictor relative to other commercially available red fluorescent dyes with maximum excitation at ~530 nm and maximum emission at ~555 nm. This product is used in a homogeneous fluorescence-based assay for detecting intracellular calcium mobilization across a broad spectrum of biological targets.
Specifications
- Purity:≥95% (HPLC)