ZnAF-2 DA
Catalog # ENZOALX620076M001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 mg
- Description:ZnAF-2 DA
- Seq:02
- Purity:≥98% (HPLC)
- Concentration:~5 mM
- Formulation:Liquid. In 0,3 ml DMSO.
Specifications
About this item
Fluorescent probe for Zn²⁺. Cell permeable.
Cell permeable derivative of ZnAF-2 (Prod. No. ALX-620-072). Fluorescent reagent (Ex(max): 492 nm; Em(max): 514 nm) for the detection of low concentration of zinc ion due to its strong affinity to zinc ion (dissociation constant: 2,7 nM). The sample zinc ion can be specifically detected. Low background fluorescense supersensitizes the visualization for in vivo sample zinc ion.