Order Entry
France
ContactUsLinkComponent
 
NUCLEAR-ID® green chromatin condensation detection kit
  ENZOENZ51021K200
 :  
NUCLEAR-ID® green chromatin condensation detection kit
  ENZOENZ51021K200
 :  

 

  • Pk:
    1 KIT
  • Description:
    NUCLEAR-ID® Green chromatin condensation detection kit
  • Seq:
    02
  • Storage conditions:
    −80 °C
  • Application:
    Flow cytometry, fluorescence microscopy, fluorescent detection
  • Size:
    1 kit
  • Tests per kit:
    200 assays

 

 

This kit provides a rapid assay for nuclear condensation, a prominent hallmark of apoptosis.A highly permeable green-emitting dye for enhanced detection of apoptosis-induced chromatin condensation.

  • Dye excitable with standard 488 nm laser
  • Intercalating dye with superior permeability with any live cell line
  • Easy no-wash mix and read protocol
  • Eliminates need for specialized 350 nm UV laser required for Hoechst dyes and reduces chances for channel interference
  • No interference from small molecule fluorescence or cell autofluorescence

The Nuclear-ID® Green Chromatin Condensation Detection Kit detects nuclear condensation using a DNA intercalating dye that brightly stains the condensed chromatin of apoptotic cells, but only dimly stains the uncondensed chromatin of healthy living cells.

The cell-permeable dye used in this application is an aromatic, planar cationic structure that inserts between stacked base pairs on the DNA duplex, providing an environmentally-dependent fluorescence enhancement of the dye molecules and large increases in fluorescence signal relative to the free dye in solution. Since the signal enhancement provides a proportional response, direct quantitation of DNA is possible.

Further signal increase is observed upon DNA condensation during apoptosis. Considering the general mutagenic effect of nucleic acid-binding dyes, careful storage and handling of this dye is recommended.

 : Kit contains Nuclear-ID™ Green Cell Cycle Detection Reagent, 100 µl Apoptosis inducer (Staurosporine), 50 nmoles 10X Assay Buffer, 30 ml.