Order Entry
France
ContactUsLinkComponent
GFP-CERTIFIED® FLUOFORTE® Calcium assay kit
GFP-CERTIFIED® FLUOFORTE® Calcium assay kit
Supplier:  ENZO LIFE SCIENCES
Certificates
GFP-CERTIFIED® FLUOFORTE® Calcium assay kit
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Storage conditions:
    –80 °C (Long term), –20 °C (Short term)
  • Application:
    Flow cytometry, fluorescence microscopy, fluorescent detection, HTS
  • Size:
    1 kit

Specifications

Frequently Bought Together