Anti-TRAIL-R3 Mouse Monoclonal Antibody [clone: DJR3]
Catalog # ENZOALX8049080100
Supplier: Enzo Life Sciences
Specifications
- Antibody type:Primary
- Antigen name:TNF Receptor Superfamily Member 10c
- Antigen symbol:TRAIL-R3
- Clonality:Monoclonal
- Clone:DJR3
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Seq:02
- Form:Liquid
- Antigen synonyms:DCR1|TRID|TRAIL-R3|TRAILR3|DCR1-TNFR|CD263|LIT
- Storage buffer:PBS, pH 7,2. containing 0,09% sodium azide.
- Storage temperature:4 °C
- Shipping temperature:Blue Ice Not Frozen
- Immunogen:Recombinant human TRAIL-R3 extracellular domain IgG1 Fc fusion protein.
- Purification:Purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
DcR1 is a 65kDa GPI-anchored member of the TNFR superfamily (TNFRSF10C) also known as TRAIL-R3, TRID, CD263, and LIT. Although not expressed by most tumors, DcR1 is expressed at low levels on normal tissues, acting as a TRAIL decoy receptor, thus inhibiting apoptosis. DcR1 is termed a decoy receptor, as it does not have a functional death domain to induce apoptosis following TRAIL ligation.
Type: Primary
Antigen: TRAIL-R3
Clonality: Monoclonal
Clone: DJR3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human