Order Entry
France
ContactUsLinkComponent
Anti-SHPS-1/SIRP-1 alpha Rabbit Polyclonal Antibody
Anti-SHPS-1/SIRP-1 alpha Rabbit Polyclonal Antibody
Catalog # ENZOADIKAPTK140E
Supplier:  ENZO LIFE SCIENCES
Anti-SHPS-1/SIRP-1 alpha Rabbit Polyclonal Antibody
Catalog # ENZOADIKAPTK140E
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    SHPS-1/SIRP-1 alpha
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Seq:
    02
  • Epitope:
    Near the C-Terminus
  • Form:
    Liquid
  • Storage buffer:
    PBS containing 0,02% sodium azide
  • Storage temperature:
    4 °C
  • Shipping temperature:
    Blue ice not frozen
  • Immunogen:
    Synthetic peptide corresponding to the sequence near the C-terminus of human SHPS-1/SIRP-1alpha
  • Purification:
    Purified by DEAE
  • Pk:
    100 µG

Specifications

About this item

SHPS-1 is a member of the gene family called the signal regulatory proteins of which there are at least fifteen members. SHPS-1 is a substrate of many activated tyrosine kinases such as Insulin receptor and EGFR, amongst others. SHPS-1 has regulatory effects on cellular responses induced by serum, growth factors, insulin, oncogenes, growth hormones and cell adhesion and plays a general role in different physiological and pathological processes.

Type: Primary
Antigen: SHPS-1/SIRP-1 alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-Terminus
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat